gRNA Design Details

Target DNA Sequence
CGCCAGGTAGAAGAGATGCG
PAM Sequence
AGG
Target locus
Chr.12: 102475755 - 102475777 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 87.72/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGF1
Entrez Gene ID
3479
Gene Name
insulin like growth factor 1
Gene Aliases
IGF-I, IGFI, MGF
Chromosome Location
Chr.12: 102395867 - 102481839 on GRCh38
UniGene ID
Hs.160562
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000618.4 NP_000609.1 2
NM_001111283.2 NP_001104753.1 2
NM_001111284.1 NP_001104754.1 2
XM_017019259.1 XP_016874748.1 2
XM_017019260.1 XP_016874749.1 2
XM_017019261.1 XP_016874750.1 2
XM_017019262.1 XP_016874751.1 2
XM_017019263.1 XP_016874752.1 2