gRNA Design Details

Target DNA Sequence
GCTTGATCCGAAACCCATCT
PAM Sequence
GGG
Target locus
Chr.3: 187285884 - 187285906 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.31/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MASP1
Entrez Gene ID
5648
Gene Name
mannan binding lectin serine peptidase 1
Gene Aliases
3MC1, CRARF, CRARF1, MAP1, MASP, MASP3, MAp44, PRSS5, RaRF
Chromosome Location
Chr.3: 187216085 - 187292220 on GRCh38
UniGene ID
Hs.89983
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001031849.2 NP_001027019.1 2
NM_001879.5 NP_001870.3 2
NM_139125.3 NP_624302.1 2
XM_006713701.2 XP_006713764.1 3
XM_011512989.2 XP_011511291.1 2
XM_011512990.2 XP_011511292.1 2
XM_011512991.2 XP_011511293.1 2
XM_017006869.1 XP_016862358.1 3
XM_017006871.1 XP_016862360.1 2
XM_017006872.1 XP_016862361.1 1