gRNA Design Details
- Target DNA Sequence
- GGTGTTTCTTGGCCCTTCGG
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 57673333 - 57673355 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRG3
- Entrez Gene ID
-
222487
- Gene Name
- adhesion G protein-coupled receptor G3
- Gene Aliases
-
GPR97, PB99, PGR26
- Chromosome Location
-
Chr.16: 57668192 - 57689378 on GRCh38
- UniGene ID
- Hs.383403
- Species
-
Human