gRNA Design Details

Target DNA Sequence
GGTGTTTCTTGGCCCTTCGG
PAM Sequence
TGG
Target locus
Chr.16: 57673333 - 57673355 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.75/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRG3
Entrez Gene ID
222487
Gene Name
adhesion G protein-coupled receptor G3
Gene Aliases
GPR97, PB99, PGR26
Chromosome Location
Chr.16: 57668192 - 57689378 on GRCh38
UniGene ID
Hs.383403
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq XM_011522953.2 XP_011521255.1 2