gRNA Design Details
- Target DNA Sequence
- TATGAAGGCGTCCATGTGTG
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 31133280 - 31133302 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.5/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS8
- Entrez Gene ID
-
5652
- Gene Name
- protease, serine 8
- Gene Aliases
-
CAP1, PROSTASIN
- Chromosome Location
-
Chr.16: 31131433 - 31135762 on GRCh38
- UniGene ID
- Hs.75799
- Species
-
Human