gRNA Design Details

Target DNA Sequence
GAAGCCTATGAGGTCAAGCT
PAM Sequence
GGG
Target locus
Chr.16: 31132918 - 31132940 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 88.53/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS8
Entrez Gene ID
5652
Gene Name
protease, serine 8
Gene Aliases
CAP1, PROSTASIN
Chromosome Location
Chr.16: 31131433 - 31135762 on GRCh38
UniGene ID
Hs.75799
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002773.4 NP_002764.1 4