gRNA Design Details
- Target DNA Sequence
- AAGTGCATCAACCACTACGG
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 65871294 - 65871316 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EFEMP2
- Entrez Gene ID
-
30008
- Gene Name
- EGF containing fibulin like extracellular matrix protein 2
- Gene Aliases
-
ARCL1B, FBLN4, MBP1, UPH1
- Chromosome Location
-
Chr.11: 65866441 - 65872934 on GRCh38
- UniGene ID
- Hs.731454
- Species
-
Human