gRNA Design Details
- Target DNA Sequence
- CGACTTTGCCTTCAGAAACT
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 46717449 - 46717471 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS50
- Entrez Gene ID
-
29122
- Gene Name
- protease, serine 50
- Gene Aliases
-
CT20, TSP50
- Chromosome Location
-
Chr.3: 46712116 - 46717883 on GRCh38
- UniGene ID
- Hs.120365
- Species
-
Human