gRNA Design Details

Target DNA Sequence
TAATGAGGCCAACCTCCAAG
PAM Sequence
TGG
Target locus
Chr.2: 157778292 - 157778314 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.67/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ACVR1
Entrez Gene ID
90
Gene Name
activin A receptor type 1
Gene Aliases
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
Chromosome Location
Chr.2: 157736446 - 157875880 on GRCh38
UniGene ID
Hs.470316
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001105.4 NP_001096.1 5
NM_001111067.2 NP_001104537.1 5
XM_005246939.3 XP_005246996.1 5
XM_005246940.3 XP_005246997.1 5
XM_006712825.3 XP_006712888.1 5
XM_011512106.2 XP_011510408.1 6
XM_011512107.2 XP_011510409.1 5
XM_011512108.2 XP_011510410.1 5