gRNA Design Details
- Target DNA Sequence
- GACTCGTTGTAGAAGCACTG
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 110942128 - 110942150 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LPAR1
- Entrez Gene ID
-
1902
- Gene Name
- lysophosphatidic acid receptor 1
- Gene Aliases
-
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
- Chromosome Location
-
Chr.9: 110873252 - 111039259 on GRCh38
- UniGene ID
- Hs.126667
- Species
-
Human