gRNA Design Details

Target DNA Sequence
GGTCTATGCTAAACAGCCTG
PAM Sequence
GGG
Target locus
Chr.15: 69360098 - 69360120 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.78/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PAQR5
Entrez Gene ID
54852
Gene Name
progestin and adipoQ receptor family member 5
Gene Aliases
MPRG
Chromosome Location
Chr.15: 69284134 - 69407637 on GRCh38
UniGene ID
Hs.591096
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001104554.1 NP_001098024.1 3
NM_017705.3 NP_060175.3 3
XM_011521720.2 XP_011520022.1 4
XM_011521722.2 XP_011520024.1 4
XM_011521725.2 XP_011520027.1 3
XM_011521727.2 XP_011520029.1 3
XM_017022360.1 XP_016877849.1 6
XM_017022361.1 XP_016877850.1 4
XM_017022362.1 XP_016877851.1 5
XM_017022363.1 XP_016877852.1 2
XM_017022364.1 XP_016877853.1 3
XM_017022365.1 XP_016877854.1 3
XM_017022366.1 XP_016877855.1 3
XM_017022367.1 XP_016877856.1 3