gRNA Design Details

Target DNA Sequence
GACCATTGCCCAGAATCCCG
PAM Sequence
AGG
Target locus
Chr.12: 108292785 - 108292807 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 89/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CMKLR1
Entrez Gene ID
1240
Gene Name
chemerin chemokine-like receptor 1
Gene Aliases
CHEMERINR, ChemR23, DEZ, RVER1
Chromosome Location
Chr.12: 108288044 - 108339348 on GRCh38
UniGene ID
Hs.197143
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001142343.1 NP_001135815.1 4
NM_001142344.1 NP_001135816.1 3
NM_001142345.1 NP_001135817.1 3
NM_004072.2 NP_004063.1 3
XM_017018820.1 XP_016874309.1 2