gRNA Design Details

Target DNA Sequence
GACTACAAGAAGAAGCGGCA
PAM Sequence
GGG
Target locus
Chr.19: 4363817 - 4363839 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.25/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SH3GL1
Entrez Gene ID
6455
Gene Name
SH3 domain containing GRB2 like 1, endophilin A2
Gene Aliases
CNSA1, EEN, SH3D2B, SH3P8
Chromosome Location
Chr.19: 4360367 - 4400568 on GRCh38
UniGene ID
Hs.490415
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001199943.1 NP_001186872.1 5
NM_001199944.1 NP_001186873.1 6
NM_003025.3 NP_003016.1 6
XM_011528183.2 XP_011526485.1 6
XM_017027146.1 XP_016882635.1 6