gRNA Design Details
- Target DNA Sequence
- CAAAGAGCCGTTACGAACAC
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 76286093 - 76286115 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
95.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRRM3
- Entrez Gene ID
-
222183
- Gene Name
- serine/arginine repetitive matrix 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 76201893 - 76287292 on GRCh38
- UniGene ID
- Hs.511025
- Species
-
Human