gRNA Design Details
- Target DNA Sequence
- GTCATGTGCGATTGTCACTC
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 34637089 - 34637111 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.24/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SIGMAR1
- Entrez Gene ID
-
10280
- Gene Name
- sigma non-opioid intracellular receptor 1
- Gene Aliases
-
ALS16, DSMA2, OPRS1, SIG-1R, SR-BP, SR-BP1, SRBP, hSigmaR1, sigma1R
- Chromosome Location
-
Chr.9: 34634722 - 34637826 on GRCh38
- UniGene ID
- Hs.522087
- Species
-
Human