gRNA Design Details
- Target DNA Sequence
- GTTTTCGTCATTGGACTTGC
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 132600548 - 132600570 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
80.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ACKR4
- Entrez Gene ID
-
51554
- Gene Name
- atypical chemokine receptor 4
- Gene Aliases
-
CC-CKR-11, CCBP2, CCR-11, CCR10, CCR11, CCRL1, CCX CKR, CCX-CKR, CKR-11, PPR1, VSHK1
- Chromosome Location
-
Chr.3: 132597237 - 132602644 on GRCh38
- UniGene ID
- Hs.310512
- Species
-
Human