gRNA Design Details

Target DNA Sequence
AAGGCGTAGGTTATGCAGCA
PAM Sequence
AGG
Target locus
Chr.20: 17514781 - 17514803 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.38/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
BFSP1
Entrez Gene ID
631
Gene Name
beaded filament structural protein 1
Gene Aliases
CP115, CP94, CTRCT33, LIFL-H
Chromosome Location
Chr.20: 17493905 - 17569220 on GRCh38
UniGene ID
Hs.129702
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001161705.1 NP_001155177.1 3
NM_001195.4 NP_001186.1 3
NM_001278606.1 NP_001265535.1 4
NM_001278607.1 NP_001265536.1 3
NM_001278608.1 NP_001265537.1 5
XM_017028005.1 XP_016883494.1 3