gRNA Design Details

Target DNA Sequence
GCCACTGAAAAGTTGTTGAG
PAM Sequence
GGG
Target locus
Chr.1: 243695593 - 243695615 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 85.32/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AKT3
Entrez Gene ID
10000
Gene Name
AKT serine/threonine kinase 3
Gene Aliases
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2
Chromosome Location
Chr.1: 243488233 - 243851079 on GRCh38
UniGene ID
Hs.498292
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001206729.1 NP_001193658.1 3
NM_005465.4 NP_005456.1 2
NM_181690.2 NP_859029.1 2
XM_005272994.4 XP_005273051.1 3
XM_005272995.2 XP_005273052.1 3
XM_006711726.3 XP_006711789.1 3
XM_011544012.2 XP_011542314.1 3
XM_011544013.2 XP_011542315.1 3
XM_016999985.1 XP_016855474.1 3