gRNA Design Details

Target DNA Sequence
TACTATTGCGCTTGCAAACA
PAM Sequence
AGG
Target locus
Chr.19: 6896463 - 6896485 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.37/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRE1
Entrez Gene ID
2015
Gene Name
adhesion G protein-coupled receptor E1
Gene Aliases
EMR1, TM7LN3
Chromosome Location
Chr.19: 6887549 - 6940453 on GRCh38
UniGene ID
Hs.2375
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001256254.1 NP_001243183.1 3