gRNA Design Details

Target DNA Sequence
CACCTTCTATCAGAATCAAG
PAM Sequence
AGG
Target locus
Chr.3: 125029411 - 125029433 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 84.07/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
HEG1
Entrez Gene ID
57493
Gene Name
heart development protein with EGF like domains 1
Gene Aliases
HEG, MST112, MSTP112
Chromosome Location
Chr.3: 124965710 - 125055958 on GRCh38
UniGene ID
Hs.477420
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_020733.1 NP_065784.1 2
XM_005247666.1 XP_005247723.1 2