gRNA Design Details

Target DNA Sequence
AAGTGTAACTCGTCTGGTGT
PAM Sequence
TGG
Target locus
Chr.5: 67100480 - 67100502 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.69/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
-
Chromosome Location
Chr.5: 66596348 - 67169595 on GRCh38
UniGene ID
Hs.595458
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001164664.1 NP_001158136.1 8
NM_001290226.1 NP_001277155.1 6
NM_001290227.1 NP_001277156.1 6
NM_001297651.1 NP_001284580.1 5
NM_015183.2 NP_055998.1 7
XM_006714606.3 XP_006714669.1 6
XM_006714610.2 XP_006714673.1 5
XM_011543382.2 XP_011541684.1 10
XM_011543384.2 XP_011541686.1 5
XM_011543385.2 XP_011541687.1 5
XM_011543386.2 XP_011541688.1 5
XM_017009447.1 XP_016864936.1 11
XM_017009448.1 XP_016864937.1 9
XM_017009449.1 XP_016864938.1 6
XM_017009450.1 XP_016864939.1 6
XM_017009451.1 XP_016864940.1 6
XM_017009452.1 XP_016864941.1 6
XM_017009453.1 XP_016864942.1 5