gRNA Design Details

Target DNA Sequence
CGACTGAGGAGGGCGTGAAT
PAM Sequence
AGG
Target locus
Chr.10: 27155473 - 27155495 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 85.85/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MASTL
Entrez Gene ID
84930
Gene Name
microtubule associated serine/threonine kinase like
Gene Aliases
GREATWALL, GW, GWL, MAST-L, THC2
Chromosome Location
Chr.10: 27154468 - 27187953 on GRCh38
UniGene ID
Hs.276905
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001172303.2 NP_001165774.1 1
NM_001172304.2 NP_001165775.1 1
NM_001320756.1 NP_001307685.1 1
NM_001320757.1 NP_001307686.1 1
NM_032844.4 NP_116233.2 1
NR_135469.1 1
XM_005252631.3 XP_005252688.1 1
XM_005252632.3 XP_005252689.1 1
XM_006717519.3 XP_006717582.1 1
XM_006717520.3 XP_006717583.1 1
XM_017016852.1 XP_016872341.1 1
XM_017016854.1 XP_016872343.1 1
XM_017016855.1 XP_016872344.1 1
XM_017016856.1 XP_016872345.1 1