gRNA Design Details
- Target DNA Sequence
- CCTGGTCGTACACCGCATAG
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 66781079 - 66781101 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
95.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SMAD6
- Entrez Gene ID
-
4091
- Gene Name
- SMAD family member 6
- Gene Aliases
-
AOVD2, HsT17432, MADH6, MADH7
- Chromosome Location
-
Chr.15: 66702110 - 66782000 on GRCh38
- UniGene ID
- Hs.153863
- Species
-
Human