gRNA Design Details

Target DNA Sequence
CCGCACCTGTGAAATCGTTG
PAM Sequence
AGG
Target locus
Chr.1: 36355409 - 36355431 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 90.44/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK40
Entrez Gene ID
83931
Gene Name
serine/threonine kinase 40
Gene Aliases
SHIK, SgK495
Chromosome Location
Chr.1: 36339619 - 36385927 on GRCh38
UniGene ID
Hs.471768
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001282546.1 NP_001269475.1 5
NM_001282547.1 NP_001269476.1 5
NM_032017.2 NP_114406.1 6