gRNA Design Details
- Target DNA Sequence
- GCCAAGCATAGTGCAGTGTT
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 36358775 - 36358797 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK40
- Entrez Gene ID
-
83931
- Gene Name
- serine/threonine kinase 40
- Gene Aliases
-
SHIK, SgK495
- Chromosome Location
-
Chr.1: 36339619 - 36385927 on GRCh38
- UniGene ID
- Hs.471768
- Species
-
Human