gRNA Design Details
- Target DNA Sequence
- CCTCAACGATTTCACAGGTG
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 36355409 - 36355431 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.04/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK40
- Entrez Gene ID
-
83931
- Gene Name
- serine/threonine kinase 40
- Gene Aliases
-
SHIK, SgK495
- Chromosome Location
-
Chr.1: 36339619 - 36385927 on GRCh38
- UniGene ID
- Hs.471768
- Species
-
Human