gRNA Design Details
- Target DNA Sequence
- GGTCGTGTCATAGTCCTCTG
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 46204275 - 46204297 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCR1
- Entrez Gene ID
-
1230
- Gene Name
- C-C motif chemokine receptor 1
- Gene Aliases
-
CD191, CKR-1, CKR1, CMKBR1, HM145, MIP1aR, SCYAR1
- Chromosome Location
-
Chr.3: 46201709 - 46208341 on GRCh38
- UniGene ID
- Hs.301921
- Species
-
Human