gRNA Design Details

Target DNA Sequence
GGTCGTGTCATAGTCCTCTG
PAM Sequence
TGG
Target locus
Chr.3: 46204275 - 46204297 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CCR1
Entrez Gene ID
1230
Gene Name
C-C motif chemokine receptor 1
Gene Aliases
CD191, CKR-1, CKR1, CMKBR1, HM145, MIP1aR, SCYAR1
Chromosome Location
Chr.3: 46201709 - 46208341 on GRCh38
UniGene ID
Hs.301921
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001295.2 NP_001286.1 2