gRNA Design Details

Target DNA Sequence
CCTGCCGCAGAACCAAGTAA
PAM Sequence
CGG
Target locus
Chr.12: 49494768 - 49494790 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SPATS2
Entrez Gene ID
65244
Gene Name
spermatogenesis associated serine rich 2
Gene Aliases
Nbla00526, P59SCR, SCR59, SPATA10
Chromosome Location
Chr.12: 49366905 - 49527441 on GRCh38
UniGene ID
Hs.654826
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001293285.1 NP_001280214.1 8
NM_001293286.1 NP_001280215.1 6
NM_023071.3 NP_075559.2 7
NR_120663.1 7
XM_011538690.2 XP_011536992.1 8
XM_011538691.2 XP_011536993.1 6
XM_017019840.1 XP_016875329.1 7