gRNA Design Details

Target DNA Sequence
CTCATGATGTACTACCAGAC
PAM Sequence
TGG
Target locus
Chr.12: 1777882 - 1777904 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.62/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADIPOR2
Entrez Gene ID
79602
Gene Name
adiponectin receptor 2
Gene Aliases
ACDCR2, PAQR2
Chromosome Location
Chr.12: 1690981 - 1788679 on GRCh38
UniGene ID
Hs.371642
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_024551.2 NP_078827.2 4
XM_005253789.1 XP_005253846.1 4
XM_006719018.2 XP_006719081.1 5
XM_011521024.2 XP_011519326.1 5