gRNA Design Details
- Target DNA Sequence
- CACAATGCCACGCTTCTGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 2159909 - 2159931 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.78/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- INS
- Entrez Gene ID
-
3630
- Gene Name
- insulin
- Gene Aliases
-
IDDM, IDDM1, IDDM2, ILPR, IRDN, MODY10
- Chromosome Location
-
Chr.11: 2159779 - 2161209 on GRCh38
- UniGene ID
- Hs.272259
- Species
-
Human