gRNA Design Details
- Target DNA Sequence
- ACCATTCGGTTCAGCATCCT
- PAM Sequence
- GGG
- Target locus
-
Chr.16: 2715847 - 2715869 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS27
- Entrez Gene ID
-
83886
- Gene Name
- protease, serine 27
- Gene Aliases
-
CAPH2, MPN
- Chromosome Location
-
Chr.16: 2712418 - 2720551 on GRCh38
- UniGene ID
- Hs.332878
- Species
-
Human