gRNA Design Details

Target DNA Sequence
TTGATGAGGACGCCCACACA
PAM Sequence
GGG
Target locus
Chr.7: 141839377 - 141839399 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 89.53/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS37
Entrez Gene ID
136242
Gene Name
protease, serine 37
Gene Aliases
TRYX2
Chromosome Location
Chr.7: 141836278 - 141849725 on GRCh38
UniGene ID
Hs.98947
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001008270.2 NP_001008271.2 2
NM_001171951.1 NP_001165422.1 2
XM_005249946.4 XP_005250003.1 2
XM_005249947.3 XP_005250004.1 3
XM_011515792.2 XP_011514094.1 3