gRNA Design Details
- Target DNA Sequence
- TCATATCCGTTCCGCACACA
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 77206969 - 77206991 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink2
- Entrez Gene ID
-
69982
- Gene Name
- serine peptidase inhibitor, Kazal type 2
- Gene Aliases
-
1700007F22Rik, AV038945, HUSI-II
- Chromosome Location
-
Chr.5: 77205106 - 77216260 on GRCm38
- UniGene ID
- Mm.46106
- Species
-
Mouse