gRNA Design Details
- Target DNA Sequence
- CAAAGACGAGCCATGCTGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 77205376 - 77205398 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.84/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink2
- Entrez Gene ID
-
69982
- Gene Name
- serine peptidase inhibitor, Kazal type 2
- Gene Aliases
-
1700007F22Rik, AV038945, HUSI-II
- Chromosome Location
-
Chr.5: 77205106 - 77216260 on GRCm38
- UniGene ID
- Mm.46106
- Species
-
Mouse