Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_152718.2 | NP_689931.2 | 4 |
XM_006718460.1 | XP_006718523.1 | 4 | |
XM_006718462.1 | XP_006718525.1 | 4 | |
XM_011544817.1 | XP_011543119.1 | 4 | |
XM_017017340.1 | XP_016872829.1 | 4 | |
XM_017017341.1 | XP_016872830.1 | 4 | |
XM_017017342.1 | XP_016872831.1 | 3 | |
XM_017017343.1 | XP_016872832.1 | 4 | |
XM_017017344.1 | XP_016872833.1 | 4 |