gRNA Design Details
- Target DNA Sequence
- GATCCCAATCCATGTGAAAA
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 84254179 - 84254201 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EDIL3
- Entrez Gene ID
-
10085
- Gene Name
- EGF like repeats and discoidin domains 3
- Gene Aliases
-
DEL1
- Chromosome Location
-
Chr.5: 83940595 - 84384867 on GRCh38
- UniGene ID
- Hs.482730
- Species
-
Human