gRNA Design Details
- Target DNA Sequence
- GTCGGAGGTCAAAGACTCTT
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 17787420 - 17787442 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SH3GL2
- Entrez Gene ID
-
6456
- Gene Name
- SH3 domain containing GRB2 like 2, endophilin A1
- Gene Aliases
-
CNSA2, EEN-B1, SH3D2A, SH3P4
- Chromosome Location
-
Chr.9: 17578954 - 17797129 on GRCh38
- UniGene ID
- Hs.743740
- Species
-
Human