gRNA Design Details
- Target DNA Sequence
- AGAAGTTCCTGACGTCAGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 74530116 - 74530138 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.32/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HTRA2
- Entrez Gene ID
-
27429
- Gene Name
- HtrA serine peptidase 2
- Gene Aliases
-
OMI, PARK13, PRSS25
- Chromosome Location
-
Chr.2: 74529405 - 74533556 on GRCh38
- UniGene ID
- Hs.469045
- Species
-
Human