gRNA Design Details
- Target DNA Sequence
- AAGCCACCTATGCACCCACT
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 133400267 - 133400289 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.87/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BFSP2
- Entrez Gene ID
-
8419
- Gene Name
- beaded filament structural protein 2
- Gene Aliases
-
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN
- Chromosome Location
-
Chr.3: 133397882 - 133475212 on GRCh38
- UniGene ID
- Hs.659862
- Species
-
Human