gRNA Design Details

Target DNA Sequence
GAAACCATTTCAGCCAACTC
PAM Sequence
AGG
Target locus
Chr.2: 196145999 - 196146021 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 86.04/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK17B
Entrez Gene ID
9262
Gene Name
serine/threonine kinase 17b
Gene Aliases
DRAK2
Chromosome Location
Chr.2: 196133583 - 196176503 on GRCh38
UniGene ID
Hs.88297
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_004226.3 NP_004217.1 4
XM_011512168.2 XP_011510470.1 4
XM_011512169.1 XP_011510471.1 4
XM_011512170.1 XP_011510472.1 5
XM_011512171.2 XP_011510473.1 3