gRNA Design Details
- Target DNA Sequence
- CCTGTTCATCAGGAACGTCG
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 89117833 - 89117855 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRSF12
- Entrez Gene ID
-
135295
- Gene Name
- serine and arginine rich splicing factor 12
- Gene Aliases
-
SFRS13B, SFRS19, SRrp35
- Chromosome Location
-
Chr.6: 89095957 - 89118081 on GRCh38
- UniGene ID
- Hs.254414
- Species
-
Human