gRNA Design Details
- Target DNA Sequence
- ACCAGCCCTCAAGTGAAAGT
- PAM Sequence
- CGG
- Target locus
-
Chr.19: 19626972 - 19626994 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.25/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LPAR2
- Entrez Gene ID
-
9170
- Gene Name
- lysophosphatidic acid receptor 2
- Gene Aliases
-
EDG-4, EDG4, LPA-2, LPA2
- Chromosome Location
-
Chr.19: 19623655 - 19628930 on GRCh38
- UniGene ID
- Hs.122575
- Species
-
Human