gRNA Design Details
- Target DNA Sequence
- TTTTCAGCGTGTCAGTGCTG
- PAM Sequence
- CGG
- Target locus
-
Chr.17: 16171862 - 16171884 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NCOR1
- Entrez Gene ID
-
9611
- Gene Name
- nuclear receptor corepressor 1
- Gene Aliases
-
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
- Chromosome Location
-
Chr.17: 16030094 - 16216746 on GRCh38
- UniGene ID
- Hs.462323
- Species
-
Human