gRNA Design Details
- Target DNA Sequence
- TGTGAAGTTACTTGAACGAG
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 8461873 - 8461895 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.79/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK33
- Entrez Gene ID
-
65975
- Gene Name
- serine/threonine kinase 33
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 8335029 - 8594289 on GRCh38
- UniGene ID
- Hs.501833
- Species
-
Human