gRNA Design Details
- Target DNA Sequence
- AGGATCTACCTCCCAAAATA
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 130207927 - 130207949 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.74/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SSMEM1
- Entrez Gene ID
-
136263
- Gene Name
- serine rich single-pass membrane protein 1
- Gene Aliases
-
C7orf45
- Chromosome Location
-
Chr.7: 130205399 - 130216869 on GRCh38
- UniGene ID
- Hs.351816
- Species
-
Human