gRNA Design Details
- Target DNA Sequence
- TGACCAGTGCCACTCTGAAG
- PAM Sequence
- CGG
- Target locus
-
Chr.16: 57563216 - 57563238 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRG5
- Entrez Gene ID
-
221188
- Gene Name
- adhesion G protein-coupled receptor G5
- Gene Aliases
-
GPR114, PGR27
- Chromosome Location
-
Chr.16: 57529339 - 57578182 on GRCh38
- UniGene ID
- Hs.187884
- Species
-
Human