gRNA Design Details
- Target DNA Sequence
- ATCAAGGCCCTAATTGGCCG
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 67909047 - 67909069 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PSKH1
- Entrez Gene ID
-
5681
- Gene Name
- protein serine kinase H1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 67893272 - 67929678 on GRCh38
- UniGene ID
- Hs.513683
- Species
-
Human