gRNA Design Details

Target DNA Sequence
TCCGGAGATCAGCACATGCT
PAM Sequence
CGG
Target locus
Chr.6: 36524434 - 36524456 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 88.68/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK38
Entrez Gene ID
11329
Gene Name
serine/threonine kinase 38
Gene Aliases
NDR, NDR1
Chromosome Location
Chr.6: 36493882 - 36547516 on GRCh38
UniGene ID
Hs.409578
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001305102.1 NP_001292031.1 4
NM_007271.3 NP_009202.1 4
XM_006714988.3 XP_006715051.1 4
XM_006714989.3 XP_006715052.1 4
XM_017010226.1 XP_016865715.1 4