gRNA Design Details
- Target DNA Sequence
- AATATCGCCATGATGTGCTG
- PAM Sequence
- GGG
- Target locus
-
Chr.14: 34435226 - 34435248 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.26/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPTSSA
- Entrez Gene ID
-
171546
- Gene Name
- serine palmitoyltransferase small subunit A
- Gene Aliases
-
C14orf147, SSSPTA
- Chromosome Location
-
Chr.14: 34432938 - 34462262 on GRCh38
- UniGene ID
- Hs.740577
- Species
-
Human